Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLC1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137756

Catalog No. 01-673-502


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is kidney or testis tissue.

Gamma Glutamylcystuene Synthetase (GSH) is the first enzyme and rate limiting step in the slutathione biosynthesis pathway. Glutathione (GSH) plays an important role in detoxification of oxidants and toxins from the cells. In most cells, GSH is synthesized denovo in two steps catalysed by Gamma Glutamylcysteine Synthetase (GCS) / Glutamate-cystein Ligase and GSH homeostasis. It is composed of two subunits: heavy and light. These subunits are coded by different genes which are controlled by different mechanisms. The heavy subunit carries the catalytic activity and can be inhibited through feedback mechanism by GSH. Binding of light subunit to heavy subunit reduces the Michaelis-Menten constant for glutamate to a physiological concentration of 1.2mM and increases the inhibitory constant for GSH.
Specifications

Specifications

GLC1D
Polyclonal
PBS with 1% BSA and <0.1% sodium azide
Liquid
GLC1D
Synthetic peptide derived from internal region of heavy subunit of GCS protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GLC1D
GLCLR, GCS light chain, GSC light chain, gamma-ECS regulatory subunit, gamma-glutamylcysteine synthetase regulatory subunit, glutamate--cysteine ligase modifier subunit, glutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase gamma-glutamylcysteine s
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2724
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit