Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526715

Catalog No. PIPA526715

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications

Specifications

GDA
Polyclonal
PBS with 0.09% sodium azide
Q9Y2T3
CYPIN, GUANASE, NEDASIN, GAH, cytoplasmic PSD95 interactor, guanine aminase, guanine aminohydrolase, p51-nedasin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9615
Flow Cytometry, Western Blot
Unconjugated
GDA
Liquid
GDA
KLH conjugated synthetic peptide between 94-122 amino acids from the N-terminal region of human GDA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit