Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-GDF-15, Polyclonal, Novus Biologicals
Manufacturer: Novus Biologicals NBP133559
Specifications
GDF-15 | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
Affinity Purified | |
GDF15 | |
Recombinant protein encompassing a sequence within the center region of human GDF15. The exact sequence is proprietary. | |
34 kDa | |
100 ul | |
Hepatic Stem Cell Markers, Stem Cell Markers | |
Primary | |
9518 | |
Human |
Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Tested in human. Not yet tested in any other species. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only