Learn More
GDI1 Rabbit anti-Bovine, Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3914
Catalog No. 01-674-577
Add to cart
Description
Description
PA3-914 detects GDI from bovine samples. PA3-914 has been successfully used in Western blot and immunofluorescence applications. The PA3-914 immunogen is purified guanine nucleotide dissociation inhibitor, isolated from bovine brain.
GDI is a protein that is used in vesicular membrane transport for recycling of Rab GTPases.
Specifications
Specifications
GDI1
Polyclonal
whole serum with 0.05% sodium azide
P21856, P31150
GDI1
Purified guanine nucleotide dissociation inhibitor, isolated from bovine brain.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2664, 281188
Immunofluorescence, Western Blot
Unconjugated
GDI1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Bovine, Human