Learn More
GDI1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA124296
Catalog No. PIPA124296
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
GDI 1 regulates the GDP/GTP exchange reaction of most RAB proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.
PA1-24296 detects GDI1 in Human samples.PA1-24296 has been successfully used in Western Blot procedures.PA1-24296 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human GDI1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GDI1
Polyclonal
Unconjugated
P31150
GDI1
Synthetic peptide region of human GDI1.
100 μg
Primary
Human
Antibody
IgG
Western Blot
1mg/mL
GDI1
XX-FW81657B9.1, FLJ41411, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
Rabbit
Protein A/G
RUO
2664
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Product Suggestions
Product Suggestions