Learn More
Gemin4 Mouse anti-Human, Clone: 17D10, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051534
Catalog No. 05-153-4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-Gemin4 Antibody, clone 17D10 (mouse monoclonal antibody) validated in western blot, immunoprecipitation to detect Gemin4 also known as HCC-associated protein 1, component of gems 4, gem (nuclear organelle) associated protein 4.
Specifications
Specifications
Gemin4
17D10
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Human
Immunoprecipitation, Western Blot
Unconjugated
NP_056536
GEMIN4, Gemin-4, HC56, HCAP1, HHRF-1, p97
Histidine-tagged recombinant protein corresponding to human Gemin4.
Protein G purfied
2-8°C for one year
Monoclonal