Learn More
Gemin5 Mouse anti-Human, Clone: 10G11, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051535
Catalog No. 05-153-5MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-Gemin5 Antibody, clone 10G11 is a Mouse Monoclonal Antibody for detection of Gemin5 also known as gem (nuclear organelle) associated protein 5 and has been validated in western blot, immunocytochemistry.
Specifications
Specifications
Gemin5
10G11
Purified Mouse Monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Purified
Murine
IgG2a, κ, κ
100μg
Primary
Human
Immunocytochemistry, Western Blot
Unconjugated
NP_056280
gemin 5, gemin5, GEMIN5, MGC142174, DKFZp586M1824
Histidine-tagged recombinant protein corresponding to human Gemin5.
Protein G purfied
2-8°C for one year
Monoclonal