Learn More
Gemin 6 Mouse anti-Human, Clone: 3D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522604
Catalog No. PIMA522604
Add to cart
Description
Description
Peptide Sequence: MSEWMKKGPL EWQDYIYKEV RVTASEKNEY KGWVLTTDPV SANIVLVNFL EDGSMSVTGI MGHAVQTVET MNEGDHRVRE KLMHLFTSGD CKAYSPEDLE ERKNSLKKWL EKNHIPITEQ GDAPRTLCVA GVLTIDPPYD PENCSSSNEI ILSRVQDLIE GHLTASQ
GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).
Specifications
Specifications
Gemin 6
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Q8WXD5
Gem (nuclear organelle) associated protein 6, FLJ23459
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79833
ELISA, Western Blot
3D2
Unconjugated
GEMIN6
Liquid
GEMIN6
GEMIN6 (AAH18195, 1 a.a. ∽ 167 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human