Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEN1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526502

Catalog No. PIPA526502


Add to cart

Description

Description

Endonuclease which cleaves flap structures at the junction between single-stranded DNA and double-stranded DNA. Specific for 5 ft.-overhanging flap structures in which the 5 ft.-upstream of the flap is completely double-stranded. Prefers the blocked-flap structures similar to those occurring at replication forks, in which the 5 ft. single-strand overhang of the flap is double-stranded.
Specifications

Specifications

GEN1
Polyclonal
PBS with 0.09% sodium azide
Q17RS7, Q8BMI4
Gen, Gen homolog 1, endonuclease, flap endonuclease GEN homolog 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
209334, 348654
Western Blot
Unconjugated
GEN1
Liquid
GEN1
KLH conjugated synthetic peptide between 60-89 amino acids from the N-terminal region of human GEN1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit