Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513817

Catalog No. PIPA513817

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1.
Specifications

Specifications

MAP4K2
Polyclonal
PBS with 0.09% sodium azide
Q12851, Q61161
Mitogen-activated protein kinase kinase kinase kinase 2; MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; Germinal center kinase; GC kinase; Rab8 interacting protein; B lymphocyte serine/threonine-protein kinase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26412, 5871
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAP4K2
Liquid
MAP4K2
KLH conjugated synthetic peptide between 288-319 amino acids from the central region of human Germinal Center Kinase (GCK/MAP4K2)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit