Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Mouse, Unlabeled, Clone: 52, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610566

 View more versions of this product

Catalog No. BDB610566


Add to cart

Description

Description

The cytoskeleton is an array of highly ordered filaments that maintains the shape, structure, and functionality of cells. There are three major types of filaments: actin filaments, microtubules, and intermediate filaments. The distinctions among them are based on protein composition and arrangements. Glial fibrillary acidic

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

GFAP
52
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.