Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Mouse anti-Human, Porcine, Clone: GFA-02, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135376

Catalog No. 01-671-431


Add to cart

Description

Description

GFAP (Glial fibrillary acidic protein) is a member of the class III intermediate filament protein family. GFAP is heavily, and specifically, expressed in astrocytes and certain astroglia in the central nervous system, in satellite cells in peripheral ganglia, and in non-myelinating Schwann cells in peripheral nerves. In addition, neural stem cells strongly express GFAP. Antibodies to GFAP are very useful as markers of astrocytic cells. In addition, many types of brain tumor, presumably derived from astrocytic cells, heavily express GFAP. GFAP is also found in the lens epithelium, Kupffer cells of the liver, in some cells in salivary tumors and has been reported in erythrocytes. GFAP is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing of the GFAP gene results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

GFAP
Monoclonal
0.1 mg/mL
PBS with 0.05% sodium azide
P14136, P03995, P47819
Glial Fibrillary Acid Protein
Mouse
IgG1
100 μg
4° C
Primary
396562, 24387, 14580, 2670
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
GFA-02
Unconjugated
GFAP
Liquid
GFAP
Porcine spinal cord.
Purified
RUO
Antibody
Monoclonal
Human, Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit