Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GFAP, Polyclonal, Abcam™

This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.

Supplier:  Abcam AB467450UL

Encompass_Preferred

Catalog No. NC9738535


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Host Species: Chicken
Isotype: IgY
Cross Reactivity: Mouse, Rat, Chicken, Cow, Cat, Human, Rhesus monkey, Apteronotus leptorhynchus. Predicted to work with all Mammals.
Immunogen: Full length native protein (purified from a Triton X-100 extract of myelin associated material, further purified by centrifugation and ion exchange chromatography in 6m urea on DEAE cellulose) (Cow).

Applications:

Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting

Specifications

Specifications

GFAP
33mg/mL
Chicken
Affinity Purified
Neuroscience, Signal Transduction, Stem Cell Biology, Tags And Cell Markers
A. leptorhynchus, Bovine, Chicken, Feline, Human, Mammalia, Mouse, Rat, Rhesus Monkey
Polyclonal
Unconjugated
Full length native protein (purified from a Triton X-100 extract of myelin associated material, further purified by centrifugation and ion exchange chromatography in 6m urea on DEAE cellulose) (Cow).
50 μL
Primary
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.