Learn More
Thermo Scientific Lab Vision GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results with Thermo Scientific GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.
Manufacturer: Thermo Scientific MS1288P1
Description
Ensure accurate, reproducible results with Thermo Scientific GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.
Host Species: Mouse
Clone: GFP01; same as B34
Isotype: IgG1/κ
Species Reactivity: Proteins from all species that are tagged with GFP.
Epitope: Not determined
Immunogen: Purified recombinant GFP protein
Molecular Weight: 27 kDa
Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins
Cellular Localization: Depends upon the localization of the parent protein tagged with GFP
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
GFP (Green Fluorescent Protein) Ab-1 | |
GFP01 (B34) | |
Unconjugated | |
Murine | |
IgG2a kappa | |
RUO | |
Primary | |
Monoclonal |
Monoclonal | |
200μg/mL | |
Concentrated | |
Purified recombinant GFP protein | |
500μL | |
Epitope Tags | |
Primary | |
All |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.