Learn More
UbpbioSupplier Diversity Partner ANTI-GFP, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1031
Antibodies – Anti-GFP (UBPBio catalog# Y1031) – Mouse monoclonal antibody (clone GF28R, IgG2b). Used for immunoblotting at 1:1000-3000 dilution for detection of 1-10 ng GFP-tagged proteins; immunostaining at 1:500-2,000 dilution; immunoprecipitation at 1-2 ug/reaction.
Catalog No. 50-112-6918