Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112532
Description
A suggested positive control for this product is human brain lysate.
GFRa-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFRa and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.Specifications
GFR alpha-1 | |
Polyclonal | |
PBS with 0.02% sodium azide | |
P56159, P97785, Q62997 | |
GFRA1 | |
Synthetic peptide corresponding to residues 369-382 of human GFR -1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
GFRA1 | |
DKFZp313E1012, DKFZp686J0156, FLJ10561, FLJ31546, GDNFR, GDNFRA, GFR-ALPHA-1, MGC23045, RET1L, RETL1, TRNR1 | |
Rabbit | |
Ion-exchange chromatography | |
RUO | |
14585, 25454, 2674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction