Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112533

Catalog No. 01-674-193


Add to cart

Description

Description

A suggested positive control for this product is HeLa whole cell lysate.

GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
Specifications

Specifications

GFR alpha-2
Polyclonal
PBS with 0.02% sodium azide
O00451, O08842
HpV18gp1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
14586, 25136, 2675
Western Blot
Unconjugated
GFRA2
Liquid
GFRA2
Synthetic peptide corresponding to residues 377-391 of human GFR -2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit