Learn More
GFRA1, Mouse, Clone: 4G3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GFRA1.
Supplier: Abnova Corporation H00002674M08
Description
Glial cell line-derived neurotrophic factor (GDNF) and neurturin (NTN) are two structurally related, potent neurotrophic factors that play key roles in the control of neuron survival and differentiation. The protein encoded by this gene is a member of the GDNF receptor family. It is a glycosylphosphatidylinositol(GPI)-linked cell surface receptor for both GDNF and NTN, and mediates activation of the RET tyrosine kinase receptor. This gene is a candidate gene for Hirschsprung disease. Multiple alternatively spliced transcript variants have been described for this gene. [provided by RefSeq
Sequence: ASDQCLKEQSCSTKYRTLRQCVAGKETNFSLASGLEAKDECRSAMEALKQKSLYNCRCKRGMKKEKNCLRIYWSMYQSLQGNDLLEDSSpecifications
GFRA1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005264 | |
GFRA1 | |
GFRA1 (NP_005255, 32 a.a. ∼ 119 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunofluorescence | |
4G3 | |
Mouse monoclonal antibody raised against a full length recombinant GFRA1. | |
GFRA1 | |
GDNFR/GDNFRA/GFR-ALPHA-1/MGC23045/RET1L/RETL1/TRNR1 | |
Mouse | |
Affinity chromatography | |
RUO | |
2674 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.