Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGH, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127920

Catalog No. PIPA127920

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Hydrolyzes the polyglutamate sidechains of pteroylpolyglutamates. Progressively removes gamma-glutamyl residues from pteroylpoly-gamma-glutamate to yield pteroyl-alpha-glutamate (folic acid) and free glutamate. May play an important role in the bioavailab

PA1-27920 detects GGH from human, mouse, and rat samples.PA1-27920 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27636 immunogen is a synthetic peptide corresponding to human Gamma-glutamyl hydrolase.
Specifications

Specifications

GGH
Polyclonal
PBS with no preservative
Q92820, Q9Z0L8, Q62867
GH
Chicken
IgY
50μg
Store at -20°C
Primary
8836
Western Blot
Unconjugated
GGH
Purified
GGH
Synthetic peptide corresponding to human Gamma-glutamyl hydrolase.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit