Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-GGT1 Clone: 1F9,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ ST1551100UG
Description
Specifically detects GGT1 Clone: 1F9 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin)
GGT1 catalyzes the transfer of the glutamyl moiety of glutathione to a variety of amino acids and dipeptide acceptors. The enzyme is composed of a heavy chain and a light chain, which are derived from a single precursor protein, and is present in tissues involved in absorption and secretion.Specifications
GGT1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
1F9 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids 381-471 of human GGT1, expressed as a GST fusion protein | |
RUO | |
Recognizes the ∽60kDa GGT1 protein in NIH3T3 cells and human testes tissue. | |
−20°C | |
IgG2 α |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction