Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127501

Catalog No. PIPA127501

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake

PA1-27501 detects GH1 from human samples.PA1-27501 may be successfully used in ELISA and Western blot applications.The immunogen is a synthetic peptide corresponding to human Somatotropin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GH1
Polyclonal
GH1
GH, GH-N, GHN, IGHD1B, hGH-N
Chicken
Antigen affinity chromatography
RUO
2688
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
ELISA, Western Blot
Unconjugated
P01241
GH1
Synthetic peptide corresponding to human Somatotropin.
50 μg
Primary
Human
Antibody
IgY
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit