Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA133080

Catalog No. PIPA133080

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake

PA1-33080 detects GH1 from human samples.PA1-33080 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-33080 immunogen is: Human growth hormone
Specifications

Specifications

GH1
Polyclonal
whole serum diluted in PBS with 1% BSA and 0.09% sodium azide
P01241
GH, GH-N, GHN, IGHD1B, hGH-N
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
GH1
Serum
GH1
Human growth hormone
6mL
Store at 4°C
Primary
2688
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit