Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHRHR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514407

Catalog No. PIPA514407

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GHRHR is a receptor for growth hormone-releasing hormone. Binding of this hormone to the receptor leads to synthesis and release of growth hormone. Mutations in GHRHR gene have been associated with isolated growth hormone deficiency (IGHD), also known as Dwarfism of Sindh, a disorder characterized by short stature.
Specifications

Specifications

GHRHR
Polyclonal
PBS with 0.09% sodium azide
Q02643
Growth hormone-releasing hormone receptor; GHRH receptor; GRF receptor; GRFR; GHRHR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2692
Western Blot
Unconjugated
GHRHR
Liquid
GHRHR
KLH conjugated synthetic peptide between 367-397 amino acids from the C-terminal region of human GHRHR
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit