Learn More
Giα1 Mouse anti-Vertebrates, Clone: 7H7, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05555
Catalog No. 05-555-MI
Add to cart
Description
Description
Anti-Giα1 Antibody, clone 7H7 detects level of Giα1 and has been published and validated for use in western blotting.
Specifications
Specifications
Giα1
7H7
0.07M Tris-glycine, pH 7.4, 0.105M NaCl, containing 30% glycerol
Purified
Murine
IgG2b
200μg
Primary
Vertebrate
Western Blot
Unconjugated
NM_002069
GNAI1, Gi
Peptide corresponding to residues 110-123 of Giα1
Protein G chromatography
−20°C for 24 months.
Monoclonal