Learn More
Giα2 Mouse anti-Rat, Bovine, Mouse, Human, Guinea Pig, Clone: L5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 051403
Catalog No. 05-140-3MI
Add to Cart
Description
Description
This Anti-Giα2 Antibody, clone L5 is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Giα2.
Specifications
Specifications
Giα2
Monoclonal
Unconjugated
NM_002070.2
Mouse
Purified
Primary
2-8°C for up to 12 months.
IgG2b
Immunohistochemistry (Paraffin), Western Blot
L5
Purified mouse monoclonal IgG2b, in 0.02Mphosphate buffer, 0.25Msodium chloride, pH 7.6, with 0.1% sodium azide.
GNAI2, GNAI2B, WUGSC:H_LUCA15.1, H_LUCA15.1, H_LUCA16.1, GIP, WUGSC:H_LUCA16.1
Recombinant Giα-2, purified by the method of Linder (J. Biol. Chem. [1990] 265, 8243).
100 μg
Bovine, Guinea Pig, Human, Mouse, Rat
Purified
Recommendations
Recommendations