Learn More
Giα3 Rabbit anti-Human, Hamster, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06270
Catalog No. 06-270-MI
Add to cart
Description
Description
Detect Giα3 using this Anti-Giα3 Antibody validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
Giα3
Unconjugated
Serum
Rabbit
IgG
50μL
Primary
NM_006496
Immunoprecipitation, Western Blot
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
GNAI3; 87U6; G; FLJ26559; ialpha-3).
peptide corresponding to the C-terminal region of Giα3 (KNNLKECGLY)
Unpurified
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Hamster, Human, Murine, Rat