Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia cysts Goat anti-Protozoa, Biotin, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173153

Catalog No. PA173153


Add to cart

Description

Description

The PA1-73153 antibody reacts with Giardia lamblia cysts. PA1-73153 has been successfully used in Immunofluorescence applications. The PA1-73153 antibody was raised against purified, intact giardia cysts. Recommended short term storage (up to 6 months) is liquid at 4°C. For longer term storage, aliquot and freeze.

Giardiasisis a diarrhoeal illness caused by a single celled microscopic protozoan parasite, Giardia lamblia, also known as Giardia intestinalis. Giardia lamblia exists in two forms, an active form called a trophozoite, and an inactive form called a cyst. The active trophozoite attaches to the lining of the small intestine and is responsible for causing the signs and symptoms of giardiasis. The trophozoite cannot live long outside of the body and spread of infection is via the cyst which is excreted in the host ft.s faeces. When it is ingested, stomach acid activates the cyst, and the cyst develops into the disease causing trophozoite in the new host. Giardiasis is diagnosed by finding cysts or trophozoites in the faeces.
Specifications

Specifications

Giardia lamblia cysts
Polyclonal
Biotin
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Protozoa
Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
G. lamblia; Giardia intestinalis
Purified, intact giardia cysts.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit