Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC3 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA132075

Catalog No. PIPA132075


Add to cart

Description

Description

PA1-32075 detects GIPC3 from human samples. PA1-32075 has been successfully used in Western blot applications. The PA1-32075 immunogen is: Peptide with sequence EGAAAREARGTET, from N Terminus of the protein sequence according to NP_573568 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GIPC3
Polyclonal
Unconjugated
GIPC3
Liquid
GIPC3
Peptide with sequence EGAAAREARGTET, from N Terminus of the protein sequence according to NP_573568
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q8TF64
C19orf64, DKFZp686J1198, FLJ40925
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
126326
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit