Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT1 Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611396

 View more versions of this product

Catalog No. BDB611396


Add to cart

Description

Description

Activation of adrenergic receptors (AR), such as β2AR leads to activation of heterotrimeric G proteins and recruitment of G protein-coupled receptor kinases (GRKs). Phosphorylation of the activated receptor by GRKs promotes the binding of arrestin protein which prevents further G protein activation. GRK interactor 1 (GIT1) was identified in a yeast two-hybrid screen for proteins that bind GRK2. GIT1 has an N-terminal Zn2+ finger-like motif involved in GTPase activating activity and multipe ankyrin repeat units. In vitro, GIT1 is a GTPase-activating protein (GAP) for the ADP ribosylation factor (ARF) family of small GTP-binding proteins. In rat, GIT1 is widely expressed with the highest levels in the testis and lowest levels in the liver and spleen. Overexpression of GIT1 in HEK293 cells causes reduced β2AR signaling and increased receptor phosphorylation as a result of reduced receptor internalization and resensitization. In addition, GIT1 interacts directly with paxillin and the PIX exchange factors, and indirectly with PAKs, Rac1, and Nck. GIT proteins are also now known to be PIP3 stimulated GAPs for ARF6. Thus, GIT1 activation of ARF proteins may involve cell signaling pathways that are important for endocytosis, cell adhesion, and cytoskeletal dynamics.

Immunofluorescence, Western Blotting

Specifications

Specifications

GIT1
13
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
G protein-coupled receptor kinase interactor 1
Affinity Purified
Rat GIT1 aa. 664-770
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.