Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT1 Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

$213.00

Specifications

Antigen GIT1
Isotype IgG2a
Immunogen Rat GIT1 aa. 664-770
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB611396
View Documents Promotion Details
BD Biosciences
611396
50μg
Each for $213.00
Add to cart
 
BDB611397
View Documents Promotion Details
BD Biosciences
611397
150μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Activation of adrenergic receptors (AR), such as β2AR leads to activation of heterotrimeric G proteins and recruitment of G protein-coupled receptor kinases (GRKs). Phosphorylation of the activated receptor by GRKs promotes the binding of arrestin protein which prevents further G protein activation. GRK interactor 1 (GIT1) was identified in a yeast two-hybrid screen for proteins that bind GRK2. GIT1 has an N-terminal Zn2+ finger-like motif involved in GTPase activating activity and multipe ankyrin repeat units. In vitro, GIT1 is a GTPase-activating protein (GAP) for the ADP ribosylation factor (ARF) family of small GTP-binding proteins. In rat, GIT1 is widely expressed with the highest levels in the testis and lowest levels in the liver and spleen. Overexpression of GIT1 in HEK293 cells causes reduced β2AR signaling and increased receptor phosphorylation as a result of reduced receptor internalization and resensitization. In addition, GIT1 interacts directly with paxillin and the PIX exchange factors, and indirectly with PAKs, Rac1, and Nck. GIT proteins are also now known to be PIP3 stimulated GAPs for ARF6. Thus, GIT1 activation of ARF proteins may involve cell signaling pathways that are important for endocytosis, cell adhesion, and cytoskeletal dynamics.

Immunofluorescence, Western Blotting

Specifications

Specifications

GIT1
Rat GIT1 aa. 664-770
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
G protein-coupled receptor kinase interactor 1
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
13
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit