Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT1 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528316

Catalog No. PIPA528316


Add to cart

Description

Description

PA5-28316 targets GIT1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-28316 immunogen is recombit fragment corresponding to a region within amino acids 375 and 631 of Human GIT1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GIT1
Polyclonal
Unconjugated
GIT1
Liquid
GIT1
Recombit fragment corresponding to a region within amino acids 375 and 631 of Human GIT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.65 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9Y2X7, Q9Z272
ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT-1, CAT1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1, cool-associated and tyrosine-phosphorylated protein 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
28964, 83709
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit