Learn More
anti-GITR Ligand, Alexa Fluor 647; Clone: MIH44; BD
Manufacturer:  BD Biosciences 563542
Catalog No. BDB563542
Add to cart
Description
Description
Applications:
Flow cytometry
Specifications
Specifications
GITR Ligand
MIH44
Alexa Fluor 647
Affinity purified
GITRL/GFP-transfected L cells
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Rat
IgG2b, kappa
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Monoclonal