Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gap junction protein, beta 5, 31.1kDa (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GJB5 protein.

Manufacturer:  Abnova Corporation H00002709B01

Catalog No. 89-005-440


Add to cart

Description

Description

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family.[supplied by OMIM

Sequence: MNWSIFEGLLSGVNKYSTAFGRIWLSLVFIFRVLVYLVTAERVWSDDHKDFDCNTRQPGCSNVCFDEFFPVSHVRLWALQLILVTCPSLLVVMHVAYREVQEKRHREAHGENSGRLYLNPGKKRGGLWWTYVCSLVFKASVDIAFLYVFHSFYPKYILPPVVKCHADPCPNIVDCFISKPSEKNIFTLFMVATAAICILLNLVELIYLVSKRCHECLAARKAQAMCTGHHPHGTTSSCKQDDLLSGDLIFLGSDSHPPLLPDRPRDHVKKTIL
Specifications

Specifications

gap junction protein, beta 5, 31.1kDa
Unconjugated
No additive
BC004379
CX31.1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2709
Western Blot
Mouse polyclonal antibody raised against a full-length human GJB5 protein.
GJB5
Serum
GJB5
GJB5 (AAH04379, 1 a.a. ~ 273 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit