Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GKAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525388

Catalog No. PIPA525388

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

GKAP1 is a protein that is highly similar to the mouse cGMP-dependent protein kinase anchoring protein 42kDa. The mouse protein has been found to localize with the Golgi and recruit cGMP-dependent protein kinase I alpha to the Golgi in mouse testes. It is thought to play a role in germ cell development. Transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

GKAP1
Polyclonal
PBS with 0.09% sodium azide
Q5VSY0
GKAP42, G kinase-anchoring protein 1, cGMP-dependent protein kinase anchoring protein 42kDa, cGMP-dependent protein kinase-anchoring protein of 42 kDa, protein kinase anchoring protein GKAP42
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80318
Immunohistochemistry, Western Blot
Unconjugated
GKAP1
Liquid
GKAP1
KLH conjugated synthetic peptide between 331-359 amino acids from the C-terminal region of human GKAP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit