Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Supplier: Novus Biologicals NBP178259SS
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In-situ Hybridization.Specifications
GLI-1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/ml | |
Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 25799059), In-situ Hybridization reported in scientific literature (PMID 24506883) | |
P08151 | |
GLI1 | |
A synthetic peptide made to an internal portion of the human Gli1 protein (between residues 150-200) [UniProt P08151] | |
0.025 mL | |
Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
2735 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction