Learn More
GLI1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532206
Description
May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.
PA5-32206 targets Gli1 in IHC (P) and WB applications and shows reactivity with Human and mouse samples.The PA5-32206 immunogen is synthetic peptide corresponding to a region within amino acids 755 and 810 of Human Gli1.
Specifications
Gli1 | |
Polyclonal | |
Unconjugated | |
P08151, P47806 | |
GLI1 | |
Synthetic peptide corresponding to a region within amino acids 755 and 810 of mouse Gli1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1.67mg/mL | |
GLI1 | |
AV235269, Zfp-5, Zfp5, glioma-associated oncogene homolog, zinc finger protein 5, zinc finger protein GLI1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14632, 2735 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.