Learn More
MilliporeSigma™ anti-Glial Fibrillary Acidic Protein (GFAP), Clone: GA5, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB3402/DEL
Catalog No. 50-175-272
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product
Description
Description
Glial fibrillary acidic protein is a class-III intermediate filament. GFAP is the main constituent of intermediate filaments in astrocytes and serves as a cell specific marker that distinguishes differentiated astrocytes from other glial cells during the development of the central nervous system.
Reference:
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Product Suggestions
Product Suggestions