anti-Glial Fibrillary Acidic Protein (GFAP), Clone: GA5, Millipore™ (Chemicon™)

Manufacturer: EMD Millipore MAB360/DEL

 View more versions of this product

Catalog No. 50-175-341

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Host Species Mouse
    Immunogen Purified GFAP from porcine spinal cord.
    Isotype IgG1
    Quantity 100μL
    Primary or Secondary Primary
    Cross Reactivity Bovine,Chicken,Human,Mouse,Porcine,Rabbit,Rat
    Antigen Glial Fibrillary Acidic Protein (GFAP)
    Format Ascites
    Regulatory Status RUO
    Research Discipline Neuroscience
    Monoclonal or Polyclonal Monoclonal
    Conjugate Unlabeled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit