Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Glis1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131273

Catalog No. NBP131273


Add to cart

Description

Description

Glis1 Polyclonal antibody specifically detects Glis1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Glis1
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
GLIS1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GLIS1. The exact sequence is proprietary.
66 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
FLJ36155, GLIS family zinc finger 1, GLI-similar 1, zinc finger protein GLIS1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
148979
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only