Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OSE00005G

Catalog No. PIOSE00005G


Add to cart

Description

Description

Reconstitute with 500 µL of distilled water.

Specifications

Specifications

GLT-1
Polyclonal
Unconjugated
SLC1A2
Lyophilized
SLC1A2
A synthetic peptide from c-terminal region of rat Excitatory amino acid transporter 2
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
P43004, P43006, P31596
Excitatory amino acid transporter 2, GLUT-R, GLT-1, Eaat2, Slc1a2, Sodium-dependent glutamate / aspartate transporter 2, Solute carrier family 1 member 2, GLT1a, GLT1b, GLT1c, solute carrier family 1 (glial high affinity glutamate transporter), member 2
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20511, 29482, 6506
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit