Learn More
GLP1R Mouse anti-Human, Clone: 4F3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB1471202
Catalog No. HYB1471202
Add to cart
Description
Description
HYB 147-12-02 detects all forms of glucogon-like peptide-1, including precursor, from human samples. HYB 147-12-02 has been successfully used in ELISA, immunohistochemistry and Western blotting procedures. HYB 147-12-02 binds to GLP-1 when coated directly onto the microtiter well, and reacts specifically with GLP-1 (7-36) amide in solution giving a Ka of 4.0x10E8 in an inhibition ELISA. HYB 147-12-02 immunogen is glucagon-like peptide-1 (7-36) amide coupled to PPD. The epitope of HYB 147-12-02 is the mid-molecular domain of GLP-1and differs from that of HYB 147-06-02 (C-terminal amide). NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
GLP-1 (7-36) amide is the principal active form of GLP-1, the other being GLP-1 (7-37). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion.
Specifications
Specifications
GLP1R
Monoclonal
1.08 mg/mL
PBS with 0.5M NaCl and 15mM sodium azide; pH 7.4
P43220
Glucagon-like peptide-1, GLP-1
Mouse
IgG1, kappa
200 μL
4° C, store in dark
Primary
2740
ELISA, Immunohistochemistry, Western Blot
4F3
Unconjugated
GLP1R
Liquid
GLP1R
Glucagon-like peptide-1 (7-36) peptide.
Protein A
RUO
Antibody
Monoclonal
Human