Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Glucagon Oligoclonal, Invitrogen™

GlucagonOligoclonal antibody 710320 from Thermo Scientific™ Pierce™ specifically detects Glucagon in Human and Mouse samples, and it is validated for WB

Manufacturer:  Invitrogen 710320

Encompass

Catalog No. 71-032-0


Add to cart

Description

Description

Glucagon is a peptide hormone secreted by alpha cells in the Islet of Langerhans of the pancreas. The peptide is synthesized as a 180 amino acid precursor which is converted to glucagon by prohormone convertase 2 (PC2). The primary role of glucagon is in gluconeogenesis and glycogenolysis. It stimulates the release of glucose by converting glycogen in liver, and also maintains glucose homeostasis in coordination with insulin.

710320 detects Glucagon in Human and Mouse samples. 710320 has been successfully used in WB procedures. The immunogen for 710320 is peptide corresponding to amino acids 98-118 of human GLP-1.

Applications:

Western Blotting

Specifications

Specifications

Glucagon
Unconjugated
P01275, P55095
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Polyclonal
Human, Murine
24HCLC
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Peptide corresponding to amino acids 98-118 of human GLP-1
Protein A
Primary
14526, 2641
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit