Learn More
Glucagon, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA185465
Catalog No. PA185465
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The protein encoded by this gene is actually a preproprotein that is cleaved into four distinct mature peptides. One of these, glucagon, is a pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. Glucagon is a ligand for a specific G-protein linked receptor whose signalling pathway controls cell proliferation. Two of the other peptides are secreted from gut endocrine cells and promote nutrient absorption through distinct mechanisms. Finally, the fourth peptide is similar to glicentin, an active enteroglucagon.
Specifications
Specifications
Glucagon
Polyclonal
Unconjugated
GCG
Liquid
GCG
Zinc glucagon-BSA.
Protein G
RUO
Antibody
Polyclonal
Human, Murine, Porcine, Rat
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays
5mg/mL
PBS with 0.09% sodium azide
P01275, P55095, P01274, P06883
GLP1, GLP2, GRPP, GLP1/2
Sheep
IgG
500μL
4°C or -20°C if preferred
Primary
14526, 24952, 2641, 397595