Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glucocorticoid Receptor, Recombinant Protein, Invitrogen™

Recombinant Protein

Manufacturer:  Invitrogen RP500

Catalog No. 01-670-216

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Glucocorticoid Receptor kRecombinant protein RP-500 detects Glucocorticoid Receptor kin Not Applicable samples, and is validated for Control, Gel Shift and WB

Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism, and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocorticoid receptor (GR) and that complex ft.s subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain glucocorticoid receptor (GR). RP-500A contains 50uL of recombinant human glucocorticoid receptor at 3.5mg/mL. RP-500B contains 20uL of oligonucleotide at 100nguL. Each RP-500 is sufficient for 50 gel shift reactions or 50 lanes for Western blot detection with any ABR polyclonal Anti-human Glucocorticoid Receptor Antibody (PA1-510A, PA1-511A, PA1-512). Gel shift and Western blot protocols are supplied with RP-500 or available upon request. The components of this kit may be used to perform gel shift assays and with PA1-510A, Anti-Glucocorticoid Receptor antibody, gel super shift assays. This complete system can be used as a control system into which the researcher may introduce other components such as other putative response elements or other glucocorticoid receptors (mutants, from various tissue sources, etc.). The recombinant protein may also be used to perform DNase I footprinting experiments, as a control in Western blot experiments or for in vitro transcription assays. RP-500A is recombinant human glucocorticoid receptor (full length) that was expressed in an insect cell expression system and enriched by ammonium sulfate precipitation followed by dialysis in KED buffer with 20% glycerol. RP-500B is an oligonucleotide double stranded DNA representing the human tyrosine amino transferase gene response element with the following sequences: 5’...CTA GGC TGT ACA GGA TGT TCT GCC TAG...3’ 3’...GAT CCG ACA TGT CCT ACA AGA CGG ATC...5’
Specifications

Specifications

Recombinant
P04150
Liquid
2908
purified
-80°C, Avoid Freeze/Thaw Cycles
GR Kit
NR3C1
Unlabeled
Insect cells
Control, Gel Shift, Western Blot
Control, Gel Shift, Western Blot
protein with no preservative
Human Glucocorticoid Receptor
1Kit
RUO
Human Glucocorticoid Receptor
Human
Hu
Recombinant
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit