Learn More
anti-Glucose 6 Phosphate Dehydrogenase, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP178074
Catalog No. NBP178074
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Glucose 6 Phosphate Dehydrogenase
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Goat
IgG
2 ml
Cancer, Cellular Markers, Golgi Apparatus Markers
Primary
2539
Bacteria
ELISA, Western Blot
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000
Delipidation and Defibrination
G6PD
Glucose-6-Phosphate Dehydrogenase [Leuconostoc mesenteroides]
Unpurified
RUO
Store at 4C. Do not freeze.
Polyclonal
This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against purified and partially purified Glucose-6-Phosphate Dehydrogenase [Leuconostoc mesenteroides]. Cross reactivity against Glucose-6-Phosphate Dehydrogenase from other tissues and species may occur but have not been specifically determined.