Learn More
Glucose-6-Phosphate Dehydrogenase Goat anti-Bacteria, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA185474
Catalog No. PA185474
Add to cart
Description
Description
Goat ani G6PDH antibody recognizes the enzyme Glucose-6-Phosphate Dehydrogenase (G6PDH).Assay by immunoelectrophoresis resulted in a single precipitin arc against anti goat serum as well as purified and partially purified glucose-6-phosphate dehydrogenase.
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
Glucose-6-Phosphate Dehydrogenase
Polyclonal
Unconjugated
Liquid
Goat
IgG
1 mL
4°C or -20°C if preferred
Primary
Bacteria
ELISA, Western Blot
5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
G6PD1
Glucose-6-Phosphate Dehydrogenase from Leuconostoc mesenteroides.
Ion-exchange chromatography
RUO
Antibody
Polyclonal