Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Glucose 6 phosphate isomerase, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP146139

Catalog No. NBP146139

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Glucose 6 phosphate isomerase
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
GPI
Peptide with sequence C-YREHRSELNLRR corresponding to internal region (near N-Terminus) according to NP_000166.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region (near N Terminus). Cross reacts with Human. Expected from sequence similarity: Pig.
ELISA, Western Blot
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:128000
NP_000166.2
AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36
Goat
IgG
0.1 mg
Cytokine Research
Primary
2821
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only