Learn More
Glucose Oxidase Mouse anti-Fungi, Clone: GO13-102.2.3.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110604
Catalog No. 01-671-557
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10604 detects Glucose Oxidase from Aspergillus niger samples. MA1-10604 has been successfully used in ELISA applications. The MA1-10604 immunogen is purified glucose oxidase from Aspergillus niger.
Specifications
Specifications
Glucose Oxidase
Monoclonal
1.46 mg/mL
PBS with no preservative; pH 7.4
GOx
Purified glucose oxidase from Aspergillus niger.
Protein G
RUO
Antibody
Monoclonal
ELISA
GO13-102.2.3.3
Unconjugated
Liquid
Mouse
IgM
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Fungi