Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP94 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137807

Catalog No. 01-673-536


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.

Heat shock protein (HSP) molecular chaperones are environmental stressinducible gene products. The human HSP 90 family includes 17 genes that fall into four classes: HSP90AA, HSP90AB, HSP90B and TRAP. HSP 90beta, also designated HSP90B, HSP 84 and HSPC2, is a cytosolic protein that participates in signaling pathways with PKC e to protect cells from external damage, particularly in heat shock-mediated events. GRP 94 (glucose regulated protein 94 kDa), also known as tumor rejection antigen 1 (TRA1), ECGP and GP96, localizes to the ER, is highly expressed in BGC-823 human gastric carcinoma cells and is upregulated in human endothelial cells in response to hypoxia by HIF-1.
Specifications

Specifications

GRP94
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.4
P14625
ECGP; GP96; GRP94; TRA1; 94 kDa glucose-regulated protein; GRP-94; Tumor rejection antigen-1; endoplasmin; endothelial cell (HBMEC) glycoprotein; glucose regulated protein; 94 kDa; gp96 homolog; heat shock protein 90 kDa beta member 1; tumor rejection antigen 1
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7184
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HSP90B1
Liquid
HSP90B1
Synthetic peptide corresponding to C-terminus of human GRP-94 protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit