Learn More
GLUT1 Mouse anti-Human, Mouse, Rat, Clone: SPM498, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA137783
Catalog No. 01672558
Add to cart
Description
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is esophageal or breast carcinoma.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein(Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon,l ung, stomach, esophagus, and breast.
Specifications
Specifications
GLUT1
Monoclonal
Unconjugated
SLC2A1
Liquid
SLC2A1
Synthetic peptide derived from C-terminus of human GLUT-1 protein
Purified
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
SPM498
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P11166, P17809, P11167
Glucose transporter type 1, erythrocyte/brain, HepG2 glucose transporter
Mouse
IgG2a, kappa
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6513, 24778, 20525